N-Phenyl-p-toluidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22038G100 AS22038G25
ACTIAS22038G100EA 0 GBP
ACTIAS22038G100 ACTIAS22038G25
N-Phenyl-p-toluidine
N-Phenyl-p-toluidine
Formula: C₁₃H₁₃N
MDL Number: MFCD00092921
CAS Number: 620-84-8
EINECS: 210-655-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR