(±)-2-Fluoro-α-methylbenzylamine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2-Fluorophenyl)ethylamine

AS20373G1 AS20373G25 AS20373G5
ACTIAS20373G1EA 0 GBP
ACTIAS20373G1 ACTIAS20373G25 ACTIAS20373G5
(±)-2-Fluoro-α-methylbenzylamine
(±)-2-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD04038307
CAS Number: 74788-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR