4'-Hydroxy-3'-methylacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25316G100 AS25316G25 AS25316G5
ACTIAS25316G100EA 0 GBP
ACTIAS25316G100 ACTIAS25316G25 ACTIAS25316G5
4'-Hydroxy-3'-methylacetophenone
4'-Hydroxy-3'-methylacetophenone
Formula: C₉H₁₀O₂
MDL Number: MFCD00002231
CAS Number: 876-02-8
EINECS: 212-880-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR