4-Methyl-3-nitroanisole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Methoxy-4-methyl-3-nitrobenzene, 4-Methoxy-2-nitrotoluene, Methyl 3-nitro-p-tolyl ether
AS21268G100
AS21268G25
AS21268G5
ACTIAS21268G100EA
319
GBP
ACTIAS21268G100
ACTIAS21268G25
ACTIAS21268G5
4-Methyl-3-nitroanisole
4-Methyl-3-nitroanisole
Formula:
C₈H₉NO₃ MW: 167.16 g/mol Boiling Pt: 266 °C (1013 hPa) Melting Pt: 17 °C Density: 1.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007173 CAS Number: 17484-36-5 EINECS: 241-500-0 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...