2-Methyl-3-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Methoxy-2-methyl-3-nitrobenzene, Methyl 3-nitro-o-tolyl ether, 2-Methoxy-6-nitrotoluene

AS20901G25 AS20901G5
ACTIAS20901G25EA 0 GBP
ACTIAS20901G25 ACTIAS20901G5
2-Methyl-3-nitroanisole
2-Methyl-3-nitroanisole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 94…96 °C (0.5 mmHg)
Melting Pt: 55…57.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007159
CAS Number: 4837-88-1
EINECS: 225-424-5
UN: 2446
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR