3-Fluoro-4-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21441G100 AS21441G25
ACTIAS21441G100EA 0 GBP
ACTIAS21441G100 ACTIAS21441G25
3-Fluoro-4-nitrotoluene
3-Fluoro-4-nitrotoluene
Formula: C₇H₆FNO₂
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.438 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007053
CAS Number: 446-34-4
EINECS: 207-166-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR