2-Fluoro-6-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20497G100 AS20497G25
ACTIAS20497G100EA 133 GBP
ACTIAS20497G100 ACTIAS20497G25
2-Fluoro-6-nitrotoluene
2-Fluoro-6-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.2766 g/cm³ (20 °C)
MDL Number: MFCD00007197
CAS Number: 769-10-8
EINECS: 212-203-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR