5-Chloro-2-nitrotoluene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22395G100
AS22395G25
ACTIAS22395G100EA
0
GBP
ACTIAS22395G100
ACTIAS22395G25
5-Chloro-2-nitrotoluene
5-Chloro-2-nitrotoluene
Formula:
C₇H₆ClNO₂ Boiling Pt: 249 °C (1013 hPa) Melting Pt: 25 °C |
MDL Number:
MFCD00024579 CAS Number: 5367-28-2 EINECS: 226-355-3 UN: 3457 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...