2-Nitrobenzyl chloride
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-(Chloromethyl)-2-nitrobenzene, 2-Nitrobenzyl chloride, α-Chloro-o-nitrotoluene, alpha-Chloro-2-nitrotoluene
AS24998G100
AS24998G25
ACTIAS24998G100EA
0
GBP
ACTIAS24998G100
ACTIAS24998G25
2-Nitrobenzyl chloride
2-Nitrobenzyl chloride
Formula:
C₇H₆ClNO₂ Boiling Pt: 263 °C (1013 hPa) Melting Pt: 49…51 °C Density: 1.5557 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007185 CAS Number: 612-23-7 EINECS: 210-300-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...