3-Hydroxy-p-toluic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Hydroxy-4-methylbenzoic acid
AS20641G100
AS20641G25
AS20641G5
ACTIAS20641G100EA
0
GBP
ACTIAS20641G100
ACTIAS20641G25
ACTIAS20641G5
3-Hydroxy-p-toluic acid
3-Hydroxy-p-toluic acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 313 Melting Pt: 207…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002511 CAS Number: 586-30-1 EINECS: 209-571-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...