3-Methyl-2-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Nitro-m-toluic acid, 3-Methyl-2-nitrobenzoic acid

AS20069G100 AS20069G25
ACTIAS20069G100EA 0 GBP
ACTIAS20069G100 ACTIAS20069G25
3-Methyl-2-nitrobenzoic acid
2-Nitro-m-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 340
Melting Pt: 218…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00007180
CAS Number: 5437-38-7
EINECS: 226-610-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR