6-Nitro-o-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methyl-6-nitrobenzoic acid

AS21016G100 AS21016G25 AS21016G5
ACTIAS21016G100EA 0 GBP
ACTIAS21016G100 ACTIAS21016G25 ACTIAS21016G5
6-Nitro-o-toluic acid
6-Nitro-o-toluic acid
Formula: C₈H₇NO₄
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 151…153 °C
MDL Number: MFCD00007267
CAS Number: 13506-76-8
EINECS: 236-833-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR