4-Amino-m-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21845G100 AS21845G25
ACTIAS21845G100EA 0 GBP
ACTIAS21845G100 ACTIAS21845G25
4-Amino-m-toluic acid
4-Amino-m-toluic acid
Formula: C₈H₉NO₂
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 170 °C
MDL Number: MFCD00007736
CAS Number: 2486-70-6
EINECS: 219-629-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR