5-Methylanthranil acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid

AS20563G100 AS20563G25
ACTIAS20563G100EA 0 GBP
ACTIAS20563G100 ACTIAS20563G25
5-Methylanthranil acid
5-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 317
Storage Temperature: Ambient
MDL Number: MFCD00007909
CAS Number: 2941-78-8
EINECS: 220-932-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR