2-Bromo-6-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

ACTIAS21179G25EA 179 GBP
ACTIAS21179G25 ACTIAS21179G100
2-Bromo-6-nitrotoluene
2-Bromo-6-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 143 °C (22 mmHg)
Melting Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009792
CAS Number: 55289-35-5
EINECS: 259-566-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR