α-Bromo-3-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20592G100 AS20592G25
ACTIAS20592G100EA 0 GBP
ACTIAS20592G100 ACTIAS20592G25
α-Bromo-3-nitrotoluene
α-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
Boiling Pt: 115…118 °C (0.8 mmHg)
Melting Pt: 59…60 °C
MDL Number: MFCD00007271
CAS Number: 3958-57-4
EINECS: 223-557-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR