4-Bromo-o-toluic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Bromo-2-methylbenzoic acid
AS21330G100
AS21330G25
AS21330G5
ACTIAS21330G100EA
0
GBP
ACTIAS21330G100
ACTIAS21330G25
ACTIAS21330G5
4-Bromo-o-toluic acid
4-Bromo-o-toluic acid
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00040905 CAS Number: 68837-59-2 EINECS: 272-437-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...