2-Chloro-5-pyridineboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21913G1 AS21913G25 AS21913G5
ACTIAS21913G1EA 0 GBP
ACTIAS21913G1 ACTIAS21913G25 ACTIAS21913G5
2-Chloro-5-pyridineboronic acid pinacol ester
2-Chloro-5-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MDL Number: MFCD03412793
CAS Number: 444120-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR