1-Aminocyclopropanecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Amino-1-cyclopropanecarboxylic acid

AS20414G100 AS20414G25 AS20414G5
ACTIAS20414G100EA 0 GBP
ACTIAS20414G100 ACTIAS20414G25 ACTIAS20414G5
1-Aminocyclopropanecarboxylic acid
1-Aminocyclopropanecarboxylic acid
Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00009944
CAS Number: 22059-21-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR