3-Hydroxyphenylboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

AS20990G1 AS20990G25 AS20990G5
ACTIAS20990G1EA 0 GBP
ACTIAS20990G1 ACTIAS20990G25 ACTIAS20990G5
3-Hydroxyphenylboronic acid pinacol ester
3-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
MDL Number: MFCD02093755
CAS Number: 214360-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR