4,5-Difluoroanthranilic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-4,5-difluorobenzoic acid

AS21605G25 AS21605G5
ACTIAS21605G25EA 0 GBP
ACTIAS21605G25 ACTIAS21605G5
4,5-Difluoroanthranilic acid
4,5-Difluoroanthranilic acid
Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
MDL Number: MFCD00077509
CAS Number: 83506-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR