4-Amino-3-(trifluoromethyl)benzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Amino-5-cyanobenzotrifluoride
AS25337G25
AS25337G1
ACTIAS25337G25EA
0
GBP
ACTIAS25337G25
ACTIAS25337G1
4-Amino-3-(trifluoromethyl)benzonitrile
4-Amino-3-(trifluoromethyl)benzonitrile
Formula:
C₈H₅F₃N₂ MW: 186.14 g/mol |
MDL Number:
MFCD00275473 CAS Number: 327-74-2 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...