3-Bromo-5-(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25345G100 AS25345G25 AS25345G5
ACTIAS25345G100EA 0 GBP
ACTIAS25345G100 ACTIAS25345G25 ACTIAS25345G5
3-Bromo-5-(trifluoromethyl)benzonitrile
3-Bromo-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MDL Number: MFCD09800708
CAS Number: 691877-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR