2-Bromo-5-(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24809G100 AS24809G25
ACTIAS24809G100EA 0 GBP
ACTIAS24809G100 ACTIAS24809G25
2-Bromo-5-(trifluoromethyl)benzonitrile
2-Bromo-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MW: 250.02 g/mol
MDL Number: MFCD03412187
CAS Number: 1483-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR