Ethyl indole-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

AS20169G100 AS20169G25
ACTIAS20169G100EA 0 GBP
ACTIAS20169G100 ACTIAS20169G25
Ethyl indole-2-carboxylate
Ethyl indole-2-carboxylate
Formula: C₁₁H₁₁NO₂
MDL Number: MFCD00005609
CAS Number: 3770-50-1
EINECS: 223-206-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR