1-Boc-3-bromoindole ≥97%

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl 3-bromo-1H-indole-1-carboxylate, tert-Butyl-3-bromo-1H-indole-1-carboxylate, 3-Bromoindole-1-carboxylic acid tert-butyl ester

AS22180G1 AS22180G25 AS22180G5
ACTIAS22180G1EA 0 GBP
ACTIAS22180G1 ACTIAS22180G25 ACTIAS22180G5
1-Boc-3-bromoindole ≥97%
1-Boc-3-bromoindole
Formula: C₁₃H₁₄BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD05864772
CAS Number: 143259-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR