6-Bromooxindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

AS21331G25 AS21331G5
ACTIAS21331G25EA 0 GBP
ACTIAS21331G25 ACTIAS21331G5
6-Bromooxindole
6-Bromooxindole
Formula: C₈H₆BrNO
MDL Number: MFCD02179605
CAS Number: 99365-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR