1-Boc-4-piperidone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20177G100
AS20177G25
ACTIAS20177G100EA
0
GBP
ACTIAS20177G100
ACTIAS20177G25
1-Boc-4-piperidone
1-Boc-4-piperidone
Formula:
C₁₀H₁₇NO₃ |
MDL Number:
MFCD00151800 CAS Number: 79099-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...