1-Boc-3-piperidone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20188G100 AS20188G25 AS20188G5
ACTIAS20188G100EA 0 GBP
ACTIAS20188G100 ACTIAS20188G25 ACTIAS20188G5
1-Boc-3-piperidone
1-Boc-3-piperidone
Formula: C₁₀H₁₇NO₃
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR