3-Fluoroanthranilic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-3-fluorobenzoic acid

AS21782G25 AS21782G5
ACTIAS21782G25EA 107 GBP
ACTIAS21782G25 ACTIAS21782G5
3-Fluoroanthranilic acid
3-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
MDL Number: MFCD01569395
CAS Number: 825-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR