3-Bromoanthranilic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Amino-3-bromobenzoic acid
AS21272G1
AS21272G25
AS21272G5
ACTIAS21272G1EA
13
GBP
ACTIAS21272G1
ACTIAS21272G25
ACTIAS21272G5
3-Bromoanthranilic acid
3-Bromoanthranilic acid
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol |
MDL Number:
MFCD03618453 CAS Number: 20776-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...