6-Bromo-m-anisaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24215G100
AS24215G25
ACTIAS24215G100EA
0
GBP
ACTIAS24215G100
ACTIAS24215G25
6-Bromo-m-anisaldehyde
6-Bromo-m-anisaldehyde
Formula:
C₈H₇BrO₂ |
MDL Number:
MFCD00463817 CAS Number: 7507-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...