5-Fluorosalicylaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20995G25 AS20995G5
ACTIAS20995G25EA 0 GBP
ACTIAS20995G25 ACTIAS20995G5
5-Fluorosalicylaldehyde
5-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
MDL Number: MFCD01090997
CAS Number: 347-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR