2-Bromo-α-methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2-Bromophenyl)ethanol

AS21665G100 AS21665G25 AS21665G5
ACTIAS21665G100EA 0 GBP
ACTIAS21665G100 ACTIAS21665G25 ACTIAS21665G5
2-Bromo-α-methylbenzyl alcohol
2-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD00065001
CAS Number: 5411-56-3
EINECS: 226-491-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR