2,6-Dinitroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS20574G100 AS20574G25 AS20574G5
ACTIAS20574G100EA 0 GBP
ACTIAS20574G100 ACTIAS20574G25 ACTIAS20574G5
2,6-Dinitroaniline
2,6-Dinitroaniline
Formula: C₆H₅N₃O₄
Melting Pt: 136 °C
Density: 1.629 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007148
CAS Number: 606-22-4
EINECS: 210-108-1
UN: 1596
ADR: 6.1,II
Merck Index: 13,03299

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR