3-Fluoro-4-nitrophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20117G25 AS20117G5
ACTIAS20117G25EA 0 GBP
ACTIAS20117G25 ACTIAS20117G5
3-Fluoro-4-nitrophenol
3-Fluoro-4-nitrophenol
Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00041251
CAS Number: 394-41-2
EINECS: 206-895-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR