2-Fluoro-4-nitrophenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20908G100
AS20908G25
ACTIAS20908G100EA
167
GBP
ACTIAS20908G100
ACTIAS20908G25
2-Fluoro-4-nitrophenol
2-Fluoro-4-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.1 g/mol Melting Pt: 118…118,5 °C |
MDL Number:
MFCD00051970 CAS Number: 403-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...