3,5-Difluorophenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22541G100
AS22541G25
ACTIAS22541G100EA
0
GBP
ACTIAS22541G100
ACTIAS22541G25
3,5-Difluorophenol
3,5-Difluorophenol
Formula:
C₆H₄F₂O MW: 130.09 g/mol Boiling Pt: 65…68 °C (15 mmHg) Melting Pt: 55 Density: 1.566 g/cm³ (20 °C) |
MDL Number:
MFCD00002255 CAS Number: 2713-34-0 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...