2,3-Difluorophenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20482G100
AS20482G25
ACTIAS20482G100EA
156
GBP
ACTIAS20482G100
ACTIAS20482G25
2,3-Difluorophenol
2,3-Difluorophenol
Formula:
C₆H₄F₂O MW: 130.09 g/mol Boiling Pt: 54 °C (25 mmHg) Melting Pt: 30…32 °C |
MDL Number:
MFCD00010262 CAS Number: 6418-38-8 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...