4-Fluoro-3-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21173G100 AS21173G25
ACTIAS21173G100EA 0 GBP
ACTIAS21173G100 ACTIAS21173G25
4-Fluoro-3-nitrobenzoic acid
4-Fluoro-3-nitrobenzoic acid
Formula: C₇H₄FNO₄
Boiling Pt: 92 °C (15 mmHg)
Melting Pt: 123…124 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR