4-Amino-3-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22680G100 AS22680G25 AS22680G5
ACTIAS22680G100EA 0 GBP
ACTIAS22680G100 ACTIAS22680G25 ACTIAS22680G5
4-Amino-3-fluorobenzoic acid
4-Amino-3-fluorobenzoic acid
Formula: C₇H₆FNO₂
MDL Number: MFCD01660374
CAS Number: 455-87-8
EINECS: 207-250-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR