5-Fluoroanthranilic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS21606G100
AS21606G25
ACTIAS21606G100EA
0
GBP
ACTIAS21606G100
ACTIAS21606G25
5-Fluoroanthranilic acid
5-Fluoroanthranilic acid
Formula:
C₇H₆FNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055566 CAS Number: 446-08-2 EINECS: 207-159-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...