4-Fluoroanthranilic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20157G100 AS20157G25 AS20157G5
ACTIAS20157G100EA 0 GBP
ACTIAS20157G100 ACTIAS20157G25 ACTIAS20157G5
4-Fluoroanthranilic acid
4-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 198…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00075553
CAS Number: 446-32-2
EINECS: 207-163-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR