2,3,5,6-Tetrafluorobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS25268G100
AS25268G25
AS25268G5
ACTIAS25268G100EA
0
GBP
ACTIAS25268G100
ACTIAS25268G25
ACTIAS25268G5
2,3,5,6-Tetrafluorobenzoic acid
2,3,5,6-Tetrafluorobenzoic acid
Formula:
C₇H₂F₄O₂ MW: 194.09 g/mol |
MDL Number:
MFCD00002408 CAS Number: 652-18-6 EINECS: 416-800-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...