3-Fluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20521G100 AS20521G25
ACTIAS20521G100EA 0 GBP
ACTIAS20521G100 ACTIAS20521G25
3-Fluorobenzonitrile
3-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1.1334 g/cm³ (25 °C)
MDL Number: MFCD00001797
CAS Number: 403-54-3
EINECS: 206-963-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR