2-Chloro-1,3-dinitrobenzene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20575G1
AS20575G25
AS20575G5
ACTIAS20575G1EA
0
GBP
ACTIAS20575G1
ACTIAS20575G25
ACTIAS20575G5
2-Chloro-1,3-dinitrobenzene
2-Chloro-1,3-dinitrobenzene
Formula:
C₆H₃ClN₂O₄ Boiling Pt: 315 °C (1013 hPa) Melting Pt: 88.5 °C Density: 1.687 g/cm³ (20 °C) |
MDL Number:
MFCD00014689 CAS Number: 606-21-3 EINECS: 210-107-6 UN: 3441 ADR: 6.1,II Merck Index: 13,02156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...