2-Chloro-4-nitrophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-2-hydroxy-5-nitrobenzene

AS22418G100 AS22418G25
ACTIAS22418G100EA 0 GBP
ACTIAS22418G100 ACTIAS22418G25
2-Chloro-4-nitrophenol
2-Chloro-4-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 109…112 °C
MDL Number: MFCD00043910
CAS Number: 619-08-9
EINECS: 210-578-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR