4-Chloro-2-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-4-methoxy-3-nitrobenzene

AS21024G100 AS21024G25
ACTIAS21024G100EA 0 GBP
ACTIAS21024G100 ACTIAS21024G25
4-Chloro-2-nitroanisole
4-Chloro-2-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 97.5
MDL Number: MFCD00024327
CAS Number: 89-21-4
EINECS: 201-887-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR