2-Chloro-4-nitroanisole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Chloro-2-methoxy-5-nitrobenzene
AS24783G100
AS24783G25
ACTIAS24783G100EA
0
GBP
ACTIAS24783G100
ACTIAS24783G25
2-Chloro-4-nitroanisole
2-Chloro-4-nitroanisole
Formula:
C₇H₆ClNO₃ MW: 187.58 g/mol |
MDL Number:
MFCD00060681 CAS Number: 4920-79-0 EINECS: 225-547-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...