3-Chloro-4-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21725G100 AS21725G25
ACTIAS21725G100EA 0 GBP
ACTIAS21725G100 ACTIAS21725G25
3-Chloro-4-fluorobenzoic acid
3-Chloro-4-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Boiling Pt: 273
Melting Pt: 136
MDL Number: MFCD00042477
CAS Number: 403-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR